Learn More
NEOGEN REVEAL Q+ FOR DON
SDP
Catalog No. NC2259325
Encompass_Preferred
Reveal® Q+ for DON. A single-step lateral flow immunochromatographic assay based on a competitive immunoassay format intended for the quantitative testing of DON in grain and grain products.
Catalog No. NC2259325 Supplier NEOGEN Supplier No. 700002539
Reveal® Q+ for DON. A single-step lateral flow immunochromatographic assay based on a competitive immunoassay format intended for the quantitative testing of DON in grain and grain products.