Learn More
TARGETMOL CHEMICALS INC Revefenacin 10MG
SDP
Catalog No. 502876269
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Revefenacin (TD-4208) is a mAChR antagonist has a high affinity on M3 receptor( Ki 0.18 nM),and potentially useful for the treatment of respiratory disease. Purity 96.66%
Catalog No. 50-287-6269 Supplier TARGETMOL CHEMICALS INC Supplier No. T739210MG
Add to Cart
Add to Cart
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Revefenacin (TD-4208) is a mAChR antagonist has a high affinity on M3 receptor( Ki 0.18 nM),and potentially useful for the treatment of respiratory disease. Purity 96.66%