Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Revefenacin | 864750-70-9 | MFCD28386316 | 99.1% | 597.75 g·mol⁻¹ | C35H43N5O4 | 10 MG
SDP

Catalog No. 5000288625
Encompass_Preferred

Revefenacin is a long-acting muscarinic acetylcholine receptor antagonist (LAMA) with high affinity for the M3 receptor (Ki = 0.18 nM). This research-grade compound is supplied as a solid (white to off-white powder) for in vitro and preclinical studies; it is for research use only and not for clinical or human use.

  • Potent M3 receptor antagonist (Ki = 0.18 nM).
  • High chemical purity (99.09%).
  • Solid, white to off-white appearance suitable for formulation or assay preparation.
  • Soluble in DMSO (≈100 mg/mL); ultrasonic agitation recommended.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.

Catalog No. 50-002-88625 Supplier Medchemexpress LLC Supplier No. HY1585110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Revefenacin is a long-acting muscarinic acetylcholine receptor antagonist (LAMA) with high affinity for the M3 receptor (Ki = 0.18 nM). This research-grade compound is supplied as a solid (white to off-white powder) for in vitro and preclinical studies; it is for research use only and not for clinical or human use.

  • Potent M3 receptor antagonist (Ki = 0.18 nM).
  • High chemical purity (99.09%).
  • Solid, white to off-white appearance suitable for formulation or assay preparation.
  • Soluble in DMSO (≈100 mg/mL); ultrasonic agitation recommended.
  • Storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C or -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.