Learn More
Enzo Life Sciences Reveromycin A (0.25mg). CAS: 134615-37-5
SDP
Supplier:  Enzo Life Sciences ALX380216MC25
Encompass_Preferred
Antibiotic. Dominant analog of the reveromycin complex. Inhibitor of the mitogenic activity of epidermal growth factor (EGF). G1 phase cell cycle inhibitor, selectively inhibiting isoleucyl-tRNA synthetase. Displays antiproliferative as well as potent antifungal activity. Induces apoptosis in osteoclasts thus inhibiting bone resorption. Formulation: Lyophilized. Purity: ≥99% (HPLC). Appearance: White to tan solid. Solubility: Soluble in ethyl acetate, 100% ethanol, methanol, dimethyl formamide or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-8980