Learn More
TARGETMOL CHEMICALS INC REVERSE T3 10MG
SDP
Catalog No. 502579180
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Reverse T3 (33'5'-triiodo-L-thyronine) inhibits the increase of sodium current generated by other thyroid hormone analogs in neonatal rat myocytes. purity: 97%
Catalog No. 50-257-9180 Supplier TARGETMOL CHEMICALS INC Supplier No. T757810MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Reverse T3 (33'5'-triiodo-L-thyronine) inhibits the increase of sodium current generated by other thyroid hormone analogs in neonatal rat myocytes. purity: 97%