Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Reverse T3 Competitive ELISA Kit

Catalog No. EEL137
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EEL137 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EEL137 Supplier Invitrogen™ Supplier No. EEL137
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The multispecies Reverse Triiodothyronine (rT3) ELISA quantitates rT3 in serum, plasma, and other biological fluids Principle of the method: The Competitive multispecies ELISA research-use-only kit is designed to quantitatively measure Reverse Triiodothyronine (rT3) independent of species. A rT3 standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a coated microtiter plate and Biotinylated Detection Ab specific to rT3 added. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) is added to each well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured on a microplate reader. The concentration of rT3 in the samples is then determined by comparing the OD of the samples to the standard curve. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Reverse triiodothyronine (rT3) is a biologically inactive form of the thyroid hormone triiodothyronine (T3). rT3 is produced by the enzymatic deiodination of T4, primarily in peripheral tissues, and is mainly found in the liver and kidneys. Its primary function is to regulate the metabolism of T3 by blocking its action. The structure of rT3 is similar to that of T3, with the exception of the position of one iodine atom. This structural difference causes rT3 to be biologically inactive and unable to bind to thyroid hormone receptors. Despite its lack of direct physiological activity, rT3 levels can be used as a marker of thyroid hormone status and metabolic function.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 93.75-6000 pg/mL
Assay Sensitivity 56.25 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Thyroid hormone
Sample Type Plasma, Serum
Target Species All Species
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Reverse triiodothyronineReverse triiodothyronine;
Interassay CV <10%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Reverse T3
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.