Learn More
Apexbio Technology LLC HyperScript Reverse Transcriptase 10000 U
SDP
Catalog No. 501998221
Encompass_Preferred
HyperScript Reverse Transcriptase is a novel reverse transcriptase engineered from M-MLV Reverse Transcriptase Compared to M-MLV Reverse Transcriptase HyperScript Reverse Transcriptase has significantly reduced RNase H activity and greatly enhanced thermostability It can withstand higher reaction temperatures and is particularly suitable for reverse transcription of RNA templates with complex secondary structures Furthermore HyperScript Reverse Transcriptase has enhanced affinity for templates making it especially suitable for reverse transcription of limiting amounts of template RNA or low-copy-number genes capable of generating cDNA products of up to 12 3 kb in length HyperScript Reverse Transcriptase is typically used for the synthesis of cDNA from RNA which can then be utilized for downstream qPCR assays or other experiments
Catalog No. 50-199-8221 Supplier Apexbio Technology LLC Supplier No. K107110000
HyperScript Reverse Transcriptase is a novel reverse transcriptase engineered from M-MLV Reverse Transcriptase Compared to M-MLV Reverse Transcriptase HyperScript Reverse Transcriptase has significantly reduced RNase H activity and greatly enhanced thermostability It can withstand higher reaction temperatures and is particularly suitable for reverse transcription of RNA templates with complex secondary structures Furthermore HyperScript Reverse Transcriptase has enhanced affinity for templates making it especially suitable for reverse transcription of limiting amounts of template RNA or low-copy-number genes capable of generating cDNA products of up to 12 3 kb in length HyperScript Reverse Transcriptase is typically used for the synthesis of cDNA from RNA which can then be utilized for downstream qPCR assays or other experiments