Learn More
INTACT GENOMICS INC igScript™ Reverse Transcriptase 50,000 Units (200 u/µl)
Supplier:  INTACT GENOMICS INC 3344
Encompass
igScript™ Reverse Transcriptase (igScript RT) is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates. igScript RT is an RNA-directed DNA polymerase which lacks 3´ → 5´ exonuclease activity and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR analysis and other applications.Applications: First strand cDNA synthesis for PCR or RT-PCR Gene expression data validation by using RT-PCRBenefits: Recombinant MMLV reverse transcriptase with greatly reduced RNase H activity. Active at temperatures up to 55°C. Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA.Product Includes: igScript™ Reverse Transcriptase 10x igScript™ RT Reaction Buffer
Catalog No. 50-000-83086