Learn More
Medchemexpress LLC Reverse transcriptase-IN-1 | 2380001-43-2 | 98.1% | 447.45 g·mol⁻¹ | C25H17N7O2 | 5 MG
SDP
Catalog No. 5000262861
Encompass_Preferred
Reverse transcriptase-IN-1 is a diarylbenzopyrimidine analogue developed as a potent, orally active nonnucleoside inhibitor of HIV-1 reverse transcriptase. Supplied as a solid for research use, the compound shows low-nanomolar antiviral activity against wild-type and common resistance-associated variants and is provided with analytical documentation and recommended storage conditions for laboratory workflows.
  • Potent nonnucleoside reverse transcriptase inhibitor.
  • Active against common resistant mutants such as K103N and E138K.
  • High reported purity (approximately 98%).
  • Solid, light yellow to yellow physical form for easy handling.
  • Stable when stored according to manufacturer recommendations (powder -20°C).
  • Supplied with datasheet, certificate of analysis, and safety data sheet for research use.
Catalog No. 50-002-62861 Supplier Medchemexpress LLC Supplier No. HY1302415MG
Reverse transcriptase-IN-1 is a diarylbenzopyrimidine analogue developed as a potent, orally active nonnucleoside inhibitor of HIV-1 reverse transcriptase. Supplied as a solid for research use, the compound shows low-nanomolar antiviral activity against wild-type and common resistance-associated variants and is provided with analytical documentation and recommended storage conditions for laboratory workflows.
  • Potent nonnucleoside reverse transcriptase inhibitor.
  • Active against common resistant mutants such as K103N and E138K.
  • High reported purity (approximately 98%).
  • Solid, light yellow to yellow physical form for easy handling.
  • Stable when stored according to manufacturer recommendations (powder -20°C).
  • Supplied with datasheet, certificate of analysis, and safety data sheet for research use.