Learn More
Medchemexpress LLC Reverse transcriptase-IN-1 | 2380001-43-2 | 98.1% | 50 MG
SDP
Catalog No. 5000339944
Encompass_Preferred
This product is a potent, orally active nonnucleoside reverse transcriptase inhibitor that acts as a diarylbenzopyrimidine analogue. It demonstrates antiviral activity against the HIV-1 reverse transcriptase enzyme, including specific HIV-1 mutants. This makes it a suitable candidate for research into human HIV-1 infection.
  • Potent inhibitory action.
  • Orally active.
  • Effective against certain viral mutants.
  • Supports research on viral infections.
Catalog No. 50-003-39944 Supplier Medchemexpress LLC Supplier No. HY13024150MG
This product is a potent, orally active nonnucleoside reverse transcriptase inhibitor that acts as a diarylbenzopyrimidine analogue. It demonstrates antiviral activity against the HIV-1 reverse transcriptase enzyme, including specific HIV-1 mutants. This makes it a suitable candidate for research into human HIV-1 infection.
  • Potent inhibitory action.
  • Orally active.
  • Effective against certain viral mutants.
  • Supports research on viral infections.