Learn More
INTACT GENOMICS INC igScript™ Glycerol-free Reverse Transcriptase 50,000 units (200 u/µl)
Catalog No. 5000082986
Encompass
igScript™ Glycerol-free Reverse Transcriptase is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates. igScript RT is an RNA-directed DNA polymerase which lacks 3´ → 5´ exonuclease activity and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR analysis and other applications.Benefits: Recombinant MMLV reverse transcriptase with greatly reduced RNase H activity. Active at temperatures up to 55°C. Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA.Product Includes: igScript™ Glycerol-free Reverse Transcriptase 10x igScript™ RT Reaction BufferStorage Temperature: –80 °C
Catalog No. 50-000-82986 Supplier INTACT GENOMICS INC Supplier No. 3344GF
igScript™ Glycerol-free Reverse Transcriptase is a recombinant Moloney Murine Leukemia Virus (MMLV) reverse transcriptase with reduced RNase H activity. It is highly efficient at producing full-length cDNA from long RNA templates. igScript RT is an RNA-directed DNA polymerase which lacks 3´ → 5´ exonuclease activity and is capable of producing cDNA from as little as 50 pg of total RNA for real-time RT-PCR analysis and other applications.Benefits: Recombinant MMLV reverse transcriptase with greatly reduced RNase H activity. Active at temperatures up to 55°C. Highly efficient at producing full-length cDNA from as little as 50 pg of total RNA.Product Includes: igScript™ Glycerol-free Reverse Transcriptase 10x igScript™ RT Reaction BufferStorage Temperature: –80 °C