Learn More
Analytical Sales & Services Reversed Phase C18 SPE 96-Well Plate, 1.0mL/100mg Bed
Catalog No. 5000806353
Encompass
C18 SPE, 1mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-06353 Supplier Analytical Sales & Services Supplier No. 9610C18100
C18 SPE, 1mL 96 Well Filter Plate, 100mg per well