Learn More
Analytical Sales & Services Reversed Phase C18 SPE 96-Well Plate, 1.0mL/25mg Bed
Catalog No. 5000806787
Encompass
C18 SPE, 1mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-06787 Supplier Analytical Sales & Services Supplier No. 9610C1825
C18 SPE, 1mL 96 Well Filter Plate, 25mg per well