Learn More
Analytical Sales & Services Reversed Phase C18 SPE 96-Well Plate, 2.0mL/100mg Bed
Catalog No. 5000805802
Encompass
C18 SPE, 2mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-05802 Supplier Analytical Sales & Services Supplier No. 9620C18100
Add to Cart
Add to Cart
C18 SPE, 2mL 96 Well Filter Plate, 100mg per well