Learn More
Analytical Sales & Services Reversed Phase C18 SPE 96-Well Plate, 2.0mL/25mg Bed
Catalog No. 5000804263
Encompass
C18 SPE, 2mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-04263 Supplier Analytical Sales & Services Supplier No. 9620C1825
C18 SPE, 2mL 96 Well Filter Plate, 25mg per well