Learn More
Analytical Sales & Services Reversed Phase C18 SPE 96-Well Plate, 2.0mL/50mg Bed
Catalog No. 5000806181
Encompass
C18 SPE, 2mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06181 Supplier Analytical Sales & Services Supplier No. 9620C1850
C18 SPE, 2mL 96 Well Filter Plate, 50mg per well