Learn More
Analytical Sales & Services Reversed Phase C2 SPE 96-Well Plate, 1.0mL/50mg Bed
Supplier:  Analytical Sales & Services 9610C250
Encompass
C2 SPE, 1mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06129