Learn More
Analytical Sales & Services Reversed Phase C8 (Encapped) SPE 96-Well Plate, 2.0mL/100mg Bed
Catalog No. 5000804259
Encompass
C8 SPE, 2mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-04259 Supplier Analytical Sales & Services Supplier No. 9620C8100
Add to Cart
Add to Cart
C8 SPE, 2mL 96 Well Filter Plate, 100mg per well