Learn More
Analytical Sales & Services Reversed Phase C8 SPE 96-Well Plate, 1.0mL/50mg Bed
Catalog No. 5000806201
Encompass
C8 SPE, 1mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06201 Supplier Analytical Sales & Services Supplier No. 9610C850
Add to Cart
Add to Cart
C8 SPE, 1mL 96 Well Filter Plate, 50mg per well