Learn More
Analytical Sales & Services Reversed Phase CH, 1.0mL/25mg
Catalog No. 5000806130
Encompass
CH (cyclohexyl) SPE, 1mL 96 Well Filter Plate, 25mg per well
Catalog No. 50-008-06130 Supplier Analytical Sales & Services Supplier No. 9610CH25
CH (cyclohexyl) SPE, 1mL 96 Well Filter Plate, 25mg per well