Learn More
Analytical Sales & Services Reversed Phase Phenyl SPE 96-Well Plate, 1.0mL/50mg Bed
Catalog No. 5000806223
Encompass
Phenyl SPE, 1mL 96 Well Filter Plate, 50mg per well
Catalog No. 50-008-06223 Supplier Analytical Sales & Services Supplier No. 9610PH50
Phenyl SPE, 1mL 96 Well Filter Plate, 50mg per well