Learn More
Analytical Sales & Services Reversed Phase Phenyl SPE 96-Well Plate, 2.0mL/100mg Bed
Catalog No. 5000806328
Encompass
Phenyl SPE, 2mL 96 Well Filter Plate, 100mg per well
Catalog No. 50-008-06328 Supplier Analytical Sales & Services Supplier No. 9620PH100
Phenyl SPE, 2mL 96 Well Filter Plate, 100mg per well