Learn More
Waters Corp Reversed-Phase QC Reference Material
Supplier:  Waters Corp 186006363
Encompass_Preferred
The Quality Control Reference Materials portfolio is a unique collection of standards and mixtures. These products allow the user to evaluate and benchmark their chromatography system before analysis of critical material. The products in the portfolio are all specially formulated based on the expertise of Waters scientist. The Reversed-Phase QC Reference Material is a significantly complex reference material mixture in that it contains compounds which could be challenging to separate. It is designed to provide this complexity and is appropriate when complex assays are routinely performed. It can be used with a variety of column chemistries and dimensions, and system hardware.
The Reversed-Phase QC Reference Material contains 7 compounds including uracil, butyl paraben, naphthalene, propranolol, dipropylphthalate, acenaphthene, and amitriptyline at pH 7.
This standard mix should be used to confirm the benchmark performance of your analytical HPLC or UPLC system.
Catalog No. 50-466-293