Learn More
Alkali Scientific Test Tube Rack, Reversible, 96x1.5/0.5mL, (112x250mm) Orange, 5/cs
SDP
Catalog No. 502335758
Encompass_Preferred
ASI's bench-top tube racks are made of 100% polypropylene. FR1505 is a reversible tube rack for 96 x 1.5mL on one side and 96 x 0.5mL on the other. Suitable for refrigerator and freezer storage as low as -80˚C. These boxes are available in orange, pack of five.
Catalog No. 50-233-5758 Supplier Alkali Scientific Supplier No. FR1505
ASI's bench-top tube racks are made of 100% polypropylene. FR1505 is a reversible tube rack for 96 x 1.5mL on one side and 96 x 0.5mL on the other. Suitable for refrigerator and freezer storage as low as -80˚C. These boxes are available in orange, pack of five.