Learn More
Mopec Reversible Backing Card Pad
SDP
Catalog No. 22446065 Shop All Mopec Products
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-446-065 White
22-446-063 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-065 Supplier Mopec Supplier No. FS285
Add to Cart
Add to Cart
Designed to mount and preserve lifted latent prints while supporting case documentation.
  • High-gloss finish on one side and matte finish on the reverse side
  • Reverse side allows writing of information related to the lifted print.
  • Made from Cast-coated cardstock with a High-gloss black or white surface for enhanced contrast.
  • Measures 0.012 in. in thickness.
Specifications
Dimensions (L x W) 3 x 5 in
Product Type Backing Card
Color White
Quantity 50/Pad