Learn More
Mopec Reversible Backing Cards
SDP
Catalog No. 22446035 Shop All Mopec Products
Change view
Click to view available options
Color:
Black
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-446-035 White
22-445-994 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-446-035 Supplier Mopec Supplier No. FS244
Add to Cart
Add to Cart
Designed to support lifted prints for documentation, display, and analysis.
  • Made from durable cast-coated cardstock.
  • Features reversible glossy and matte sides.
Specifications
Product Type Backing Card
Color White
Quantity 100/Pk.