Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reversible Graph It! Mat
SCI_ED

Catalog No. S79407
Change view
Click to view available options
Grade:
K to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S79407 K to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79407 Supplier Learning Resources, Inc Supplier No. LER 0215

missing translation for 'validMainframeMsg'

Teach graphing using a 4 x 12 grid and a Venn Diagram with this double-sided vinyl mat. Measures 24 x 72 in. (60 x 180cm).

Order Info

Grades PK-3

Specifications

Grade K to 12
Format Hands-on Learning
Product Type Reversible Graph It! Mats
Subject Mathematics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.