Learn More
Electron Microscopy Sciences SEM Mount Holder Box Universal
SDP
Catalog No. 5029562
Encompass_Preferred
SEM specimen mount holder box with insert and lid holds 12 mounts.
Use: 10 or 15 mm cylinders or 3.2 mm pins
Catalog No. 50-295-62 Supplier Electron Microscopy Sciences Supplier No. 76510
SEM specimen mount holder box with insert and lid holds 12 mounts.
Use: 10 or 15 mm cylinders or 3.2 mm pins