Learn More
Alkali Scientific ASI™ Reversible Rack for 60 x 1.5/2.0mL or 0.5mL Microtubes, Blue, 5/Pack
SDP
Catalog No. 503026871
Encompass_Preferred
These reversible racks are designed to keep 0.5mL and 1.5mL( up to 2.0mL) microcentrifuge tubes organized and upright. Made of polypropylene, choose the blue rack for 60 tube capacity (5 rows of 12 tubes) racks have alphanumeric markings and are supplied in packs of 5. Reversible rack Blue color Holds 60 x 1.5/2.0mL or 0.5mL microtubes 5 racks per pack Accept 0.5 and 1.5 to 2.0mL microtubes
Catalog No. 50-302-6871 Supplier Alkali Scientific Supplier No. FR220B
These reversible racks are designed to keep 0.5mL and 1.5mL( up to 2.0mL) microcentrifuge tubes organized and upright. Made of polypropylene, choose the blue rack for 60 tube capacity (5 rows of 12 tubes) racks have alphanumeric markings and are supplied in packs of 5. Reversible rack Blue color Holds 60 x 1.5/2.0mL or 0.5mL microtubes 5 racks per pack Accept 0.5 and 1.5 to 2.0mL microtubes