Learn More
Thomas Scientific Reversible Reagent Reservoir, PP
SDP
Catalog No. NC0940208
Encompass
This versatile polypropylene reservoir is useful for single and multichannel micropipettes. Positioned one way. It provides a 50 mL reservoir for use with a single channel micropipette. Flipped over, it provides a numbered reservoir with eight separate 5 mL reagent channels, and a numbered reservoir with twelve separate 5 mL reagent channels. Autoclavable.
Catalog No. NC0940208 Supplier Thomas Scientific Supplier No. 1192T70
This versatile polypropylene reservoir is useful for single and multichannel micropipettes. Positioned one way. It provides a 50 mL reservoir for use with a single channel micropipette. Flipped over, it provides a numbered reservoir with eight separate 5 mL reagent channels, and a numbered reservoir with twelve separate 5 mL reagent channels. Autoclavable.