Learn More
Apexbio Technology LLC Reversine 656820-32-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064365
Encompass_Preferred
Reversine (CAS 656820-32-5) is a small molecule inhibitor targeting Aurora kinases a family of serine/threonine kinases involved in mitotic progression Reversine inhibits Aurora kinase A B and C with IC50 values of 150 nM 500 nM and 400 nM respectively In vitro studies show reversine can induce dedifferentiation of murine myoblast cells and possesses regenerative potential Additionally reversine displays anti-tumor effects by suppressing the expression of Aurora kinases thus disrupting cell-cycle progression In cervical carcinoma mouse models (U14 cells) reversine combined with aspirin synergistically decreases tumor growth highlighting reversine s value for cancer biology research
Catalog No. 50-000-64365 Supplier Apexbio Technology LLC Supplier No. A37605.1
Reversine (CAS 656820-32-5) is a small molecule inhibitor targeting Aurora kinases a family of serine/threonine kinases involved in mitotic progression Reversine inhibits Aurora kinase A B and C with IC50 values of 150 nM 500 nM and 400 nM respectively In vitro studies show reversine can induce dedifferentiation of murine myoblast cells and possesses regenerative potential Additionally reversine displays anti-tumor effects by suppressing the expression of Aurora kinases thus disrupting cell-cycle progression In cervical carcinoma mouse models (U14 cells) reversine combined with aspirin synergistically decreases tumor growth highlighting reversine s value for cancer biology research