Learn More
TARGETMOL CHEMICALS INC REVERSINE 25MG
SDP
Catalog No. 502581085
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Reversine a small synthetic purine analogue (2 6-disubstituted purine) is a potent inhibitior of Aurora A/B/C(IC50s=150-500 nM). purity: 99%
Catalog No. 50-258-1085 Supplier TARGETMOL CHEMICALS INC Supplier No. T182525MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Reversine a small synthetic purine analogue (2 6-disubstituted purine) is a potent inhibitior of Aurora A/B/C(IC50s=150-500 nM). purity: 99%