Learn More
Abcam Reversine, Cell-permeable A3 receptor antagonist, 5MG
Catalog No. 502707131
Encompass_Preferred
MW 393.49 g/mol, Purity >98%. Reversine, Cell-permeable A3 receptor antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7131 Supplier Abcam Supplier No. AB1209215MG
MW 393.49 g/mol, Purity >98%. Reversine, Cell-permeable A3 receptor antagonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement