Learn More
LICORbio Revert™ 520 Total Protein Stain Kits for Western Blot Normalization (500 mL kit)
Catalog No. NC1992333
Encompass_Preferred
Make Western blot normalization more accurate and reliable with Revert 520 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 520 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 500 mL Kit is enough for 50 mini blots and contains 1 x 500 mL Revert 520 Total Protein Stain, 1 x 1 L Revert 520 Wash Solution, and 1 x 1 L Revert Destaining Solution
Catalog No. NC1992333 Supplier LICORbio Supplier No. 92610016
Make Western blot normalization more accurate and reliable with Revert 520 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 520 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 500 mL Kit is enough for 50 mini blots and contains 1 x 500 mL Revert 520 Total Protein Stain, 1 x 1 L Revert 520 Wash Solution, and 1 x 1 L Revert Destaining Solution