Learn More
LICORbio Revert™ 520 Total Protein Stain and Wash Solution Kit (1 Kit)
Catalog No. NC1992332
Encompass_Preferred
Revert 520 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 520 nm channel for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes. This kit contains 200 mL Revert 520 Total Protein Stain and 400 mL Revert 520 Wash Solution. This kit is sufficient for 20 mini Western blots.
Catalog No. NC1992332 Supplier LICORbio Supplier No. 92610015
Revert 520 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 520 nm channel for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes. This kit contains 200 mL Revert 520 Total Protein Stain and 400 mL Revert 520 Wash Solution. This kit is sufficient for 20 mini Western blots.