Learn More
LICORbio Revert™ 520 Total Protein Stain for Western Blot Normalization (200 mL)
Catalog No. NC1992329
Encompass_Preferred
Make Western blot normalization more accurate and reliable with Revert 520 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 520 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. Revert 520 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 520 nm channel for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes. The 200 mL size is sufficient for 20 mini Western blots (5 mL/blot), and the 500 mL size is sufficient for 50 mini Western blots.
Catalog No. NC1992329 Supplier LICORbio Supplier No. 92610011
Make Western blot normalization more accurate and reliable with Revert 520 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 520 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. Revert 520 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 520 nm channel for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes. The 200 mL size is sufficient for 20 mini Western blots (5 mL/blot), and the 500 mL size is sufficient for 50 mini Western blots.