Learn More
LICORbio Revert™ 520 Total Protein Stain Kits for Western Blot Normalization (200 mL kit)
Catalog No. NC1992328
Encompass_Preferred
Make Western blot normalization more accurate and reliable with Revert 520 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 520 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 200 mL Kit is enough for 20 mini blots and contains 1 x 200 mL Revert 520 Total Protein Stain, 1 x 400 mL Revert 520 Wash Solution, and 1 x 400 mL Revert Destaining Solution
Catalog No. NC1992328 Supplier LICORbio Supplier No. 92610010
Make Western blot normalization more accurate and reliable with Revert 520 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 520 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 200 mL Kit is enough for 20 mini blots and contains 1 x 200 mL Revert 520 Total Protein Stain, 1 x 400 mL Revert 520 Wash Solution, and 1 x 400 mL Revert Destaining Solution