Learn More
LICORbio Revert™ 700 Total Protein Stain and Wash Solution Kit (1 kit)
Catalog No. NC1097476
Encompass_Preferred
Revert 700 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 700nm channel of Odyssey Imaging Systems for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes. This kit contains 100 mL Revert 700 Total Protein Stain and 200 mL Revert 700 Wash Solution. This kit is sufficient for 20 mini Western blots.
Catalog No. NC1097476 Supplier LICORbio Supplier No. 92611015
Revert 700 Total Protein Stain is a robust and replicable solution for Western blot normalization. The stain can be detected in the 700nm channel of Odyssey Imaging Systems for quantification and normalization analyses. The protocol is quick and can be completed within 15 minutes. This kit contains 100 mL Revert 700 Total Protein Stain and 200 mL Revert 700 Wash Solution. This kit is sufficient for 20 mini Western blots.