Learn More
LICORbio Revert™ 700 Total Protein Stain Kits for Western Blot Normalization (250 mL kit)
Catalog No. NC1572646
Encompass_Preferred
Make Western blot normalization more accurate and reliable with Revert 700 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 700 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 250 mL Kit is enough for 50 mini blots and contains 1 x 250 mL Revert 700 Total Protein Stain, 1 x 500 mL Revert 700 Wash Solution, and 1 x 500 mL Revert Destaining Solution
Catalog No. NC1572646 Supplier LICORbio Supplier No. 92611016
Make Western blot normalization more accurate and reliable with Revert 700 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 700 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 250 mL Kit is enough for 50 mini blots and contains 1 x 250 mL Revert 700 Total Protein Stain, 1 x 500 mL Revert 700 Wash Solution, and 1 x 500 mL Revert Destaining Solution