Learn More
LICORbio Revert™ 700 Total Protein Stain Kits for Western Blot Normalization (100 mL kit)
Catalog No. NC1145693
Encompass_Preferred
Make Western blot normalization more accurate and reliable with Revert 700 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 700 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 100 mL Kit is enough for 20 mini blots and contains 1 x 100 mL Revert 700 Total Protein Stain, 1 x 200 mL Revert 700 Wash Solution, and 1 x 200 mL Revert Destaining Solution
Catalog No. NC1145693 Supplier LICORbio Supplier No. 92611010
Make Western blot normalization more accurate and reliable with Revert 700 Total Protein Stain, a membrane-based (post-transfer) normalization strategy that stains all protein in your sample. Revert 700 Total Protein Stain works with your existing Western blot workflow and does not require special gels or equipment. The 100 mL Kit is enough for 20 mini blots and contains 1 x 100 mL Revert 700 Total Protein Stain, 1 x 200 mL Revert 700 Wash Solution, and 1 x 200 mL Revert Destaining Solution