Learn More
Rockland Immunochemicals Revitablot™ Western Blot Stripping Buffer - MB-085-0500
SDP
Catalog No. 501059041
Encompass_Preferred
Revitablot™ Western Blot Stripping Buffer is a uniquely formulated, ready-to-use reagent specifically designed for western blotting. Revitablot™ Western Blot Stripping Buffer is faster and more efficient at stripping of primary and secondary antibodies, including HRP and fluorescent labeled. Blots can be stripped multiple times for the repeated use of membranes. This solution can be used at room temperature. Only incubation of 5-20 minutes needed to strip the membrane. Revitablot™ Western Blot Stripping Buffer is gentle and effective on both nitrocellulose and PVDF western blot membranes.
Catalog No. 50-105-9041 Supplier Rockland Immunochemicals Supplier No. MB0850500
Revitablot™ Western Blot Stripping Buffer is a uniquely formulated, ready-to-use reagent specifically designed for western blotting. Revitablot™ Western Blot Stripping Buffer is faster and more efficient at stripping of primary and secondary antibodies, including HRP and fluorescent labeled. Blots can be stripped multiple times for the repeated use of membranes. This solution can be used at room temperature. Only incubation of 5-20 minutes needed to strip the membrane. Revitablot™ Western Blot Stripping Buffer is gentle and effective on both nitrocellulose and PVDF western blot membranes.