Learn More
Biocare Medical, LLC Revival Series Sampler
SDP
Catalog No. NC1632353
Encompass_Preferred
Revival Series Sampler was designed as a diluent kit to test for optimal staining of primary antibodies. These diluents are specifically formulated to enhance primary antibody staining and may increase titers up to 2 to 4 times.The color-coded Revival Series - Da Vinci Green, Monet Blue ,Van Gogh Yellow and Renoir Red diluents were compared to the majority of PBS-based and Tris-based diluents that are available on the market. In the vast majority of cases, either Da Vinci Green, Monet Blue, Van Gogh Yellow or Renoir Red diluents demonstrated improved staining. These diluents can also be used on an automated staining system.
Catalog No. NC1632353 Supplier Biocare Medical, LLC Supplier No. PD912H4
Revival Series Sampler was designed as a diluent kit to test for optimal staining of primary antibodies. These diluents are specifically formulated to enhance primary antibody staining and may increase titers up to 2 to 4 times.The color-coded Revival Series - Da Vinci Green, Monet Blue ,Van Gogh Yellow and Renoir Red diluents were compared to the majority of PBS-based and Tris-based diluents that are available on the market. In the vast majority of cases, either Da Vinci Green, Monet Blue, Van Gogh Yellow or Renoir Red diluents demonstrated improved staining. These diluents can also be used on an automated staining system.