Learn More
Meridian Bioscience™ Revogene™ GBS LB
GSA_VA
Catalog No. 23029053 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
24
Catalog No. Quantity
23-029-053 24
1 options
This item is not returnable. View return policy
Catalog No. 23-029-053 Supplier Meridian Bioscience™ Supplier No. 410200
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Precise diagnostic test for detection of Group B Streptococcus (GBS) from Lim Broth samples.
  • Efficient workflow
  • Flexible platform
  • Platform for growth
  • Total solution
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Content And Storage 2°C to 25°C
Type Group B (Streptococcus)
Includes One Sample Buffer Tube (SBT); One Disposable Transfer Tool (DTT); One GBS LB PIE
Quantity 24