Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Revogene™ with ImmunoCard STAT! Strep A
GSA_VA

Catalog No. 23029055PM Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity: 192 Revogene Strep A tests and 150 ImmunoCard STAT! Strep A tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
23-029-055PM Quantity: 192 Revogene Strep A tests and 150 ImmunoCard STAT! Strep A tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-055PM Supplier Meridian Bioscience™ Supplier No. 410400P43
Add to Cart
Add to Cart
This item is not returnable. View return policy

Differentiate Group A Strep from viral phayrngitis by screening with ImmunoCard STAT! Strep A and confirming all negative rapid tests with Revogene Strep A.

  • Provide a near patient solution to meet your testing needs
  • A rapid, clia-waived test for the detection of Group A Streptococcal antigen from throat swabs
  • If positive, physicians can treat
  • If negative, they can confirm with Revogene Strep A
  • Confirmation with Revogene Strep A can help prevent unnecessary antibiotic treatment by delivering accurate, actionable results for faster treatement decisions and improved outcomes

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Content And Storage ImmunoCard STAT! Strep A: 15 to 30°C; Revogene Strep A: 2 to 25°C
CE Marker Yes
Detectable Analytes Streptococci (Group A), S. pyogenes Nucleic Acids
Format Immunocard
Type Immunochromatographic Lateral Flow, PCR
For Use With (Application) Revogene
Includes 4 Revogene Strep A kits and 3 ImmunoCard STAT! Strep A kits
Quantity 192 Revogene Strep A tests and 150 ImmunoCard STAT! Strep A tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.