Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meridian Bioscience™ Revogene™ Strep A

Catalog No. 23029055 Shop All Meridian Bioscience Products
Change view
Click to view available options
Quantity:
24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-029-055 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-029-055 Supplier Meridian Bioscience™ Supplier No. 410400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Precise diagnostic test for detection of Group A Streptococcus (GAS) from throat swab specimens.

  • Efficient workflow
  • Flexible platform
  • Platform for growth
  • Total solution

Specifications

Content And Storage 2°C to 25°C
Type Group A
Includes One Sample Buffer Tube (SBT); One Disposable Transfer Tool (DTT); One GBS LB PIE
Quantity 24
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.