Learn More
Med Vet International Revolution Plum For Dogs, 85.1-130 Lbs, 3-Pack
Catalog No. 502828702
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Revolution (selamectin) Topical Parasiticide kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of American dog tick (Dermacentor variabilis) infestations in dogs. For use in dogs 6 weeks and older.
Catalog No. 50-282-8702 Supplier Med Vet International Supplier No. RXREVPLUM
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Revolution (selamectin) Topical Parasiticide kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of American dog tick (Dermacentor variabilis) infestations in dogs. For use in dogs 6 weeks and older.