Learn More
Med Vet International Zoetis Revolution Purple For Dogs, 5.1-10 Lbs, 6-Pack
Catalog No. 502827990
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Revolution (selamectin) Topical Parasiticide kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of American dog tick (Dermacentor variabilis) infestations in dogs. For use in dogs 6 weeks and older.
Catalog No. 50-282-7990 Supplier Med Vet International Supplier No. RXREVPUR
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Revolution (selamectin) Topical Parasiticide kills adult fleas and prevents flea eggs from hatching for one month and is indicated for the prevention and control of flea infestations (Ctenocephalides felis), prevention of heartworm disease caused by Dirofilaria immitis, and the treatment and control of ear mite (Otodectes cynotis) infestations. Also is indicated for the treatment and control of sarcoptic mange (Sarcoptes scabiei) and for the control of American dog tick (Dermacentor variabilis) infestations in dogs. For use in dogs 6 weeks and older.