Learn More
Grainger KNIPEX Revolving Punch Plier, 8-3/10"
Catalog No. NC1884241
Encompass_Preferred
The KNIPEX Revolving Punch Plier is an 8-inch plier designed for punching clean holes into soft materials like leather, plastic, rubber, and soft metals up to 3mm thick. It has a revolving head with 6 interchangeable dies for making holes from 2-10mm in diameter. The tool is drop-forged from chrome vanadium electric steel and has an ergonomic, non-slip plastic coating on the handles. It can punch up to 2000 holes per charge. The plier weighs 0.75 lbs.
Catalog No. NC1884241 Supplier Grainger Supplier No. 10U132
The KNIPEX Revolving Punch Plier is an 8-inch plier designed for punching clean holes into soft materials like leather, plastic, rubber, and soft metals up to 3mm thick. It has a revolving head with 6 interchangeable dies for making holes from 2-10mm in diameter. The tool is drop-forged from chrome vanadium electric steel and has an ergonomic, non-slip plastic coating on the handles. It can punch up to 2000 holes per charge. The plier weighs 0.75 lbs.