Learn More
Medchemexpress LLC Revumenib | 2169919-21-3 | 99.99% | 630.82 | 200 MG
SDP
Catalog No. 5000349992
Encompass_Preferred
Revumenib (SNDX-5613) is a potent and specific Menin-MLL inhibitor with a binding Ki of 0.149 nM and a cell-based IC50 of 10-20 nM. It is intended for the research of MLL-rearranged (MLL-r) acute leukemias, including acute lymphoblastic leukemia (ALL) and acute myeloid leukemia (AML). For research use only; not for sale to patients.
  • Potent and specific Menin-MLL inhibitor.
  • Applicable for research in MLL-rearranged acute leukemias (ALL and AML).
  • Shows in vivo plasma IC50 of 53 nM.
  • Provides significant survival benefit and leukemic control in aggressive MOLM-13 disseminated xenografts in vivo.
Catalog No. 50-003-49992 Supplier Medchemexpress LLC Supplier No. HY136175200MG
Revumenib (SNDX-5613) is a potent and specific Menin-MLL inhibitor with a binding Ki of 0.149 nM and a cell-based IC50 of 10-20 nM. It is intended for the research of MLL-rearranged (MLL-r) acute leukemias, including acute lymphoblastic leukemia (ALL) and acute myeloid leukemia (AML). For research use only; not for sale to patients.
  • Potent and specific Menin-MLL inhibitor.
  • Applicable for research in MLL-rearranged acute leukemias (ALL and AML).
  • Shows in vivo plasma IC50 of 53 nM.
  • Provides significant survival benefit and leukemic control in aggressive MOLM-13 disseminated xenografts in vivo.