Learn More
Selleck Chemical LLC Revumenib-5mg
SDP
Catalog No. 502389640
Encompass_Preferred
Revumenib(SNDX-5613) is a potent and selective inhibitor of menin-MLL binding with a Ki of 0.15 nM. SNDX-5613 shows anti-proliferative activity against multiple cell lines harboring MLLr translocations (MV4;11, RS4;11, MOLM-13, KOPN-8) with IC50 values ranging from 10-20 nM.
Catalog No. 50-238-9640 Supplier Selleck Chemical LLC Supplier No. S89965MG
Revumenib(SNDX-5613) is a potent and selective inhibitor of menin-MLL binding with a Ki of 0.15 nM. SNDX-5613 shows anti-proliferative activity against multiple cell lines harboring MLLr translocations (MV4;11, RS4;11, MOLM-13, KOPN-8) with IC50 values ranging from 10-20 nM.